L-Glutaminsäure-5-benzylester ≥99%
Lieferant: Thermo Scientific
Synonyme:
γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH
B22184.06
B22184.14
B22184.06EA
76.3
EUR
B22184.06
B22184.14
L-Glutaminsäure-5-benzylester ≥99%
L-Glutaminsäure-5-benzylester
Purity: 99%
CAS No.: 1676-73-9
Molecular Formula: C12H15NO4
Formula Weight: 237.26
Storage Temperature: 2°C to 8°C
Melting Point: 162-165°
Beilstein Reference: 1885646
Optical Rotation: +23° (c=1 in acetic acid)
MDL No.: MFCD00002633
Formel:
C₁₂H₁₅NO₄ Molecular Weight: 237,3 g/mol Schmelzpunkt: 162…165 °C |
MDL:
MFCD00002633 CAS-Nummer: 1676-73-9 EINECS: 216-826-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...