L-Glutaminsäure-5-benzylester

Lieferant: BACHEM BIOCHEMICA

Synonyme: γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH

4000146.0005 4000146.0025 4000146.0100
BACE4000146.0005EA 0 EUR
BACE4000146.0005 BACE4000146.0025 BACE4000146.0100
L-Glutaminsäure-5-benzylester
L-Glutaminsäure-5-benzylester
Formel: C₁₂H₁₅NO₄
Molecular Weight: 237,3 g/mol
MDL: MFCD00002633
CAS-Nummer: 1676-73-9
EINECS: 216-826-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR