L-Glutaminsäure-5-benzylester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH
49510-25G
49510-5G
SIAL49510-25GEA
95.2
EUR
SIAL49510-25G
SIAL49510-5G
L-Glutaminsäure-5-benzylester, Sigma-Aldrich®
L-Glutaminsäure-5-benzylester
Formel:
C₁₂H₁₅NO₄ Molecular Weight: 237,3 g/mol |
MDL:
MFCD00002633 CAS-Nummer: 1676-73-9 EINECS: 216-826-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...