1-Benzyl-4,4-difluorpiperidin

Lieferant: FLUOROCHEM

013121-1G 013121-5G 013121-25G
FLUO013121-1GEA 34.1 EUR
FLUO013121-1G FLUO013121-5G FLUO013121-25G
1-Benzyl-4,4-difluorpiperidin
1-Benzyl-4,4-difluorpiperidin
Formel: C₁₂H₁₅F₂N
MDL: MFCD03840201
CAS-Nummer: 155137-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR