6,8-Dibromo-3-formylchromone

Lieferant: CHEMPUR

Synonyme: 6,8-Dibromo-4-oxo-4H-1-benzopyran-3-carboxaldehyde

FL12011.POR 012011-5G
CHMPFL12011.POREA 0 EUR
CHMPFL12011.POR FLUO012011-5G
6,8-Dibromo-3-formylchromone
6,8-Dibromo-3-formylchromone
Formel: C₁₀H₄Br₂O₃
Molecular Weight: 331,945 g/mol
MDL: MFCD00191991
CAS-Nummer: 42059-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR