6-Chlorchromon-2-carbonsäure

Lieferant: CHEMPUR

FL12035.POR 012035-1G 012035-5G
CHMPFL12035.POREA 0 EUR
CHMPFL12035.POR FLUO012035-1G FLUO012035-5G
6-Chlorchromon-2-carbonsäure
6-Chlorchromon-2-carbonsäure
Formel: C₁₀H₅ClO₄
MDL: MFCD00847039
CAS-Nummer: 5006-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR