3-Formyl-6-methylchromon
Lieferant: CHEMPUR
Synonyme:
6-Methyl-4-oxo-4H-chromene-3-carbaldehyde
FL12002.POR
012002-10G
012002-25G
CHMPFL12002.POREA
0
EUR
CHMPFL12002.POR
FLUO012002-10G
FLUO012002-25G
3-Formyl-6-methylchromon
3-Formyl-6-methylchromon
Formel:
C₁₁H₈O₃ Molecular Weight: 188,1794 g/mol |
MDL:
MFCD00138943 CAS-Nummer: 42059-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...