6-Methylchromon-2-carbonsäure

Lieferant: CHEMPUR

FL12034.POR 012034-5G
CHMPFL12034.POREA 0 EUR
CHMPFL12034.POR FLUO012034-5G
6-Methylchromon-2-carbonsäure
6-Methylchromon-2-carbonsäure
Formel: C₁₁H₈O₄
MDL: MFCD00239435
CAS-Nummer: 5006-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR