Diphenylphosphinoxid 97%

Lieferant: Thermo Scientific
Achtung

310120010 310120050
ACRO310120010EA 56.8 EUR
ACRO310120010 ACRO310120050
Diphenylphosphinoxid 97%
Diphenylphosphine oxide
Formel: C₁₂H₁₁OP
Molecular Weight: 202,20 g/mol
Schmelzpunkt: 50…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002079
CAS-Nummer: 4559-70-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 50 to 57 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR