Ethyl-3-amino-1H-pyrazol-4-carboxylat
Lieferant: FLUOROCHEM
Synonyme:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
005978-1G
005978-5G
FLUO005978-1GEA
31.1
EUR
FLUO005978-1G
FLUO005978-5G
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel:
C₆H₉N₃O₂ |
MDL:
MFCD00005238 CAS-Nummer: 6994-25-8 EINECS: 230-262-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...