1H-Pyrazol-3-carbonsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid
B25732.03
B25732.06
B25732.03EA
46.4
EUR
B25732.03
B25732.06
1H-Pyrazol-3-carbonsäure ≥97%
1H-Pyrazole-3-carboxylic acid
Purity: 97%
CAS No.: 1621-91-6
Molecular Formula: C4H4N2O2
Formula Weight: 112.09
Melting Point: 211° dec.
MDL No.: MFCD00077436
Formel:
C₄H₄N₂O₂ Molecular Weight: 112,09 g/mol Schmelzpunkt: 211 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077436 CAS-Nummer: 1621-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...