1H-Pyrazol-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid

014533-1G 014533-5G
FLUO014533-1GEA 31.1 EUR
FLUO014533-1G FLUO014533-5G
1H-Pyrazol-3-carbonsäure
1H-Pyrazole-3-carboxylic acid
Formel: C₄H₄N₂O₂
Molecular Weight: 112.09 g/mol
MDL: MFCD00077436
CAS-Nummer: 1621-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR