4-Iod-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iodopyrazole

HC-6322-5G HC-6322-25G HC-6322-100G
COBBHC-6322-5GEA 25.5 EUR
COBBHC-6322-5G COBBHC-6322-25G COBBHC-6322-100G
4-Iod-1H-pyrazol
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193,97 g/mol
MDL: MFCD00005244
CAS-Nummer: 3469-69-0
EINECS: 222-434-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR