4-Iod-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 4-Iodopyrazole

005458-5G 005458-25G
FLUO005458-5GEA 31.1 EUR
FLUO005458-5G FLUO005458-25G
4-Iod-1H-pyrazol
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.97 g/mol
MDL: MFCD00005244
CAS-Nummer: 3469-69-0
EINECS: 222-434-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR