1-Phenyl-1H-pyrazol-4-carbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde

HI-1708-5G HI-1708-1G
COBBHI-1708-5GEA 259 EUR
COBBHI-1708-5G COBBHI-1708-1G
1-Phenyl-1H-pyrazol-4-carbaldehyd
1-Phenyl-1H-pyrazol-4-carbaldehyd
Formel: C₁₀H₈N₂O
Molecular Weight: 172,18 g/mol
MDL: MFCD02179568
CAS-Nummer: 54605-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR