3-Amino-5-phenyl-1H-pyrazol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Amino-5-phenylpyrazole

A12701.06 A12701.14
A12701.06EA 111 EUR
A12701.06 A12701.14
3-Amino-5-phenyl-1H-pyrazol 98%
3-Amino-5-phenyl-1H-pyrazole

Purity: 98% CAS No.: 1572-10-7 Molecular Formula: C9H9N3 Formula Weight: 159.19 Melting Point: 124-128° Beilstein Reference: 4947

Formel: C₉H₉N₃
Molecular Weight: 159.19 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191749
CAS-Nummer: 1572-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR