5-Brom-4-(2,5-dimethylphenyl)pyrimidin

Lieferant: COMBI-BLOCKS

HC-6109-5G HC-6109-1G
COBBHC-6109-5GEA 149 EUR
COBBHC-6109-5G COBBHC-6109-1G
5-Brom-4-(2,5-dimethylphenyl)pyrimidin
5-Bromo-4-(2,5-dimethylphenyl)pyrimidine
Formel: C₁₂H₁₁BrN₂
MDL: MFCD09475896
CAS-Nummer: 941294-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR