5-Brom-4-o-tolylpyrimidin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-4-(2-methylphenyl)pyrimidine
HC-6108-5G
HC-6108-1G
COBBHC-6108-5GEA
375
EUR
COBBHC-6108-5G
COBBHC-6108-1G
5-Brom-4-o-tolylpyrimidin
5-Brom-4-o-tolylpyrimidin
Formel:
C₁₂H₁₁BrN₂ |
MDL:
MFCD09475895 CAS-Nummer: 941294-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...