5-Brom-4-(3,4-dimethylphenyl)pyrimidin

Lieferant: COMBI-BLOCKS

HC-6121-5G HC-6121-1G
COBBHC-6121-5GEA 149 EUR
COBBHC-6121-5G COBBHC-6121-1G
5-Brom-4-(3,4-dimethylphenyl)pyrimidin
5-Bromo-4-(3,4-dimethylphenyl)pyrimidine
Formel: C₁₂H₁₁BrN₂
MDL: MFCD09800943
CAS-Nummer: 941294-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR