5-Brom-4-p-tolylpyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-4-(4-methylphenyl)pyrimidine

HC-6127-1G HC-6127-5G
COBBHC-6127-1GEA 49.7 EUR
COBBHC-6127-1G COBBHC-6127-5G
5-Brom-4-p-tolylpyrimidin
5-Brom-4-p-tolylpyrimidin
Formel: C₁₂H₁₁BrN₂
MDL: MFCD09800948
CAS-Nummer: 149323-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR