2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril

Lieferant: BIONET RESEARCH

6Y-0837.10 6Y-0837.5 6Y-0837.1 6Y-0837.500
BIOT6Y-0837.10EA 0 EUR
BIOT6Y-0837.10 BIOT6Y-0837.5 BIOT6Y-0837.1 BIOT6Y-0837.500
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
Formel: C₁₁H₇N₄Br
MDL: MFCD06659571
CAS-Nummer: 749885-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR