2-Benzyloxy-5-brompyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Benzyloxy)-5-bromopyrimidine

PY-7963-1G PY-7963-5G
COBBPY-7963-1GEA 98.3 EUR
COBBPY-7963-1G COBBPY-7963-5G
2-Benzyloxy-5-brompyrimidin
2-Benzyloxy-5-brompyrimidin
Formel: C₁₁H₉BrN₂O
Molecular Weight: 265,11 g/mol
MDL: MFCD07375140
CAS-Nummer: 742058-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR