1,2-Epoxycyclopentan ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyclopentene oxide

B20629.06 B20629.14
B20629.06EA 59.4 EUR
B20629.06 B20629.14
1,2-Epoxycyclopentan ≥97%
1,2-Epoxycyclopentane

Purity: 97% CAS No.: 285-67-6 Molecular Formula: C5H8O Formula Weight: 84.12 Storage Temperature: 2°C to 8°C Flash Point: 10°(50°F) Refractive Index: 1.4340 Boiling Point: 101-102° Beilstein Reference: 102495 UN No.: 1993 MDL No.: MFCD00005161

Formel: C₅H₈O
Molecular Weight: 84,12 g/mol
Siedepunkt: 101…102 °C
Dichte: 0,965
Flammpunkt: 10 °C (50 °F)
MDL: MFCD00005161
CAS-Nummer: 285-67-6
EINECS: 206-005-6
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR