2-Acetamido-5-bromthiazol

Lieferant: COMBI-BLOCKS

Synonyme: N-(5-bromothiazol-2-yl)acetamide

HC-6857-25G HC-6857-1G HC-6857-5G
COBBHC-6857-25GEA 1050 EUR
COBBHC-6857-25G COBBHC-6857-1G COBBHC-6857-5G
2-Acetamido-5-bromthiazol
2-Acetamido-5-bromthiazol
Formel: C₅H₅BrN₂OS
Molecular Weight: 221,0800 g/mol
MDL: MFCD00017336
CAS-Nummer: 7336-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR