2-Acetamidothiazol

Lieferant: COMBI-BLOCKS

HI-1258-25G HI-1258-100G HI-1258-5G
COBBHI-1258-25GEA 186 EUR
COBBHI-1258-25G COBBHI-1258-100G COBBHI-1258-5G
2-Acetamidothiazol
2-Acetamidothiazol
Formel: C₅H₆N₂OS
Molecular Weight: 142,18 g/mol
MDL: MFCD00016892
CAS-Nummer: 2719-23-5
EINECS: 220-321-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR