(2-Chlor-1,3-thiazol-5-yl)methylacetat

Lieferant: BIONET RESEARCH

7N-088.500 7N-088.1 7N-088.5 7N-088.10
BIOT7N-088.500EA 0 EUR
BIOT7N-088.500 BIOT7N-088.1 BIOT7N-088.5 BIOT7N-088.10
(2-Chlor-1,3-thiazol-5-yl)methylacetat
(2-Chlor-1,3-thiazol-5-yl)methylacetat
Formel: C₆H₆NO₂ClS
MDL: MFCD01443869
CAS-Nummer: 339018-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR