Ethyl-2-amino-4-methylthiazol-5-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Ethyl 2-amino-4-methylthiazole-5-carboxylate

8M-594S.500 8M-594S.1 8M-594S.5 8M-594S.10
BIOT8M-594S.500EA 0 EUR
BIOT8M-594S.500 BIOT8M-594S.1 BIOT8M-594S.5 BIOT8M-594S.10
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186,22 g/mol
MDL: MFCD00123414
CAS-Nummer: 7210-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR