Ethyl-2-amino-4-methylthiazol-5-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-amino-4-methylthiazole-5-carboxylate

HC-6710-5G HC-6710-25G HC-6710-100G
COBBHC-6710-5GEA 28.5 EUR
COBBHC-6710-5G COBBHC-6710-25G COBBHC-6710-100G
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186,22 g/mol
MDL: MFCD00123414
CAS-Nummer: 7210-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR