Ethyl-2,4-dimethylthiazol-5-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2,4-dimethylthiazole-5-carboxylate
HC-6758-5G
HC-6758-25G
HC-6758-1G
COBBHC-6758-5GEA
238
EUR
COBBHC-6758-5G
COBBHC-6758-25G
COBBHC-6758-1G
Ethyl-2,4-dimethylthiazol-5-carboxylat
Ethyl-2,4-dimethylthiazol-5-carboxylat
Formel:
C₈H₁₁NO₂S Molecular Weight: 185.24 g/mol |
MDL:
MFCD00052874 CAS-Nummer: 7210-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...