1,2:7,8-Diepoxyoctan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4-Di(oxiran-2-yl)butane

M-Clarity™ Qualitätsstufe = MQ200

139564-10G 139564-50G
SIAL139564-10GEA 74.8 EUR
SIAL139564-10G SIAL139564-50G
1,2:7,8-Diepoxyoctan, Sigma-Aldrich®
1,2:7,8-Diepoxyoctane
Formel: C₈H₁₄O₂
Molecular Weight: 142,20 g/mol
MDL: MFCD00005155
CAS-Nummer: 2426-07-5
EINECS: 219-375-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR