Phenylglycidylether ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Glycidyl phenyl ether
B21768.0B
B21768.18
B21768.30
B21768.0BEA
385
EUR
B21768.0B
B21768.18
B21768.30
Phenylglycidylether ≥99%
Phenylglycidylether
Purity: 99%
CAS No.: 122-60-1
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 114°(237°F)
Refractive Index: 1.5310
Odour: Sweet
Melting Point: 3.5°
Boiling Point: 244-246°
Beilstein Reference: 2744
MDL No.: MFCD00005133
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Siedepunkt: 244…246 °C Schmelzpunkt: 3,5 °C Dichte: 1,110 Flammpunkt: 114 °C (237 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005133 CAS-Nummer: 122-60-1 EINECS: 204-557-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...