L-Glutaminsäure-5-mthylester
Lieferant: BACHEM BIOCHEMICA
Synonyme:
5-Ethyl L-Glutamate, L-Glutamic acid 5-Ethyl ester, H-Glu(OEt)-OH
4014122.0025
4014122.0100
BACE4014122.0025EA
0
EUR
BACE4014122.0025
BACE4014122.0100
L-Glutaminsäure-5-mthylester
L-Glutaminsäure-5-mthylester
Formel:
C₇H₁₃NO₄ Molecular Weight: 175.2 g/mol |
MDL:
MFCD00037217 CAS-Nummer: 1119-33-1 EINECS: 214-274-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...