L-Glutaminsäure-5-methylester ≥99%
Lieferant: Thermo Scientific
Synonyme:
H-Glu(OMe)-OH
B24859.06
B24859.14
B24859.22
B24859.06EA
23.8
EUR
B24859.06
B24859.14
B24859.22
L-Glutaminsäure-5-methylester ≥99%
L-Glutaminsäure-5-methylester
Purity: 99%
CAS No.: 1499-55-4
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Storage Temperature: 2°C to 8°C
Melting Point: 182° dec.
MDL No.: MFCD00002632
Formel:
C₆H₁₁NO₄ Molecular Weight: 161,2 g/mol Schmelzpunkt: 182 °C (Zersetzung) |
MDL:
MFCD00002632 CAS-Nummer: 1499-55-4 EINECS: 216-110-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...