L(+)-Penicillamin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

416870010 416870050
ACRO416870010EA 47.2 EUR
ACRO416870010 ACRO416870050
L(+)-Penicillamin 98%
L(+)-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.21 g/mol
Schmelzpunkt: 206 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00064303
CAS-Nummer: 1113-41-3
EINECS: 214-203-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation +61° to +65° (20°C, 589 nm) (c=1, 1 M NaOH)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR