L(+)-Penicillamin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

4031087.0005 4031087.0025
BACE4031087.0005EA 0 EUR
BACE4031087.0005 BACE4031087.0025
L(+)-Penicillamin
L(+)-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.21 g/mol
MDL: MFCD00064303
CAS-Nummer: 1113-41-3
EINECS: 214-203-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR