L(+)-Penicillamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
M-Clarity™ Qualitätsstufe = MQ100
196312-1G
196312-5G
SIAL196312-1GEA
61
EUR
SIAL196312-1G
SIAL196312-5G
L(+)-Penicillamin, Sigma-Aldrich®
L(+)-Penicillamin
Formel:
C₅H₁₁NO₂S Molecular Weight: 149.21 g/mol |
MDL:
MFCD00064303 CAS-Nummer: 1113-41-3 EINECS: 214-203-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...