4-Acetamido-2-brom-5-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1578-5G AN-1578-1G
COBBAN-1578-5GEA 884 EUR
COBBAN-1578-5G COBBAN-1578-1G
4-Acetamido-2-brom-5-nitroanisol
4-Acetamido-2-brom-5-nitroanisol
Formel: C₉H₉BrN₂O₄
MDL: MFCD16618927
CAS-Nummer: 7357-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR