Trimethylsilylbromacetat

Lieferant: CHEMPUR SILANE

SILS19100.10 SILS19100.25
SILNSILS19100.10EA 0 EUR
SILNSILS19100.10 SILNSILS19100.25
Trimethylsilylbromacetat
Trimethylsilylbromacetat
Formel: C₅H₁₁BrO₂Si
MDL: MFCD00000187
CAS-Nummer: 18291-80-0
EINECS: 242-165-3
UN: 2920
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR