4'-Brom-2'-(trifluormethoxy)acetanilid
Lieferant: Thermo Scientific
TL00670DA
TL00670.10
TL00670.50
MAYBTL00670DAEA
0
EUR
MAYBTL00670DA
MAYBTL00670.10
MAYBTL00670.50
4'-Brom-2'-(trifluormethoxy)acetanilid
4'-Brom-2'-(trifluormethoxy)acetanilid
Formel:
C₉H₇BrF₃NO₂ |
MDL:
MFCD00203979 CAS-Nummer: 175278-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...