2-Cyan-3',4'-dichloracetanilid

Lieferant: BIONET RESEARCH

Synonyme: 2-Cyano-N-(3,4-dichlorophenyl)acetamide

2D-095.500 2D-095.1 2D-095.5 2D-095.10
BIOT2D-095.500EA 0 EUR
BIOT2D-095.500 BIOT2D-095.1 BIOT2D-095.5 BIOT2D-095.10
2-Cyan-3',4'-dichloracetanilid
2-Cyan-3',4'-dichloracetanilid
Formel: C₉H₆Cl₂N₂O
MDL: MFCD00036014
CAS-Nummer: 15386-80-8
EINECS: 239-410-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR