3'-Chlor-2-cyanacetanilid

Lieferant: SIA ENAMINE

EN300-01397.10 EN30001397.1G
ENAMEN300-01397.10EA 0 EUR
ENAMEN300-01397.10 ENAMEN30001397.1G
3'-Chlor-2-cyanacetanilid
3'-Chlor-2-cyanacetanilid
Formel: C₉H₇ClN₂O
CAS-Nummer: 17722-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR