4'-Chlor-2-cyanacetanilid

Lieferant: BIONET RESEARCH

Synonyme: N-(4-Chlorophenyl)-2-cyanoacetamide

9B-047.500 9B-047.1 9B-047.5 9B-047.10
BIOT9B-047.500EA 0 EUR
BIOT9B-047.500 BIOT9B-047.1 BIOT9B-047.5 BIOT9B-047.10
4'-Chlor-2-cyanacetanilid
4'-Chlor-2-cyanacetanilid
Formel: C₉H₇ClN₂O
Molecular Weight: 194.6177 g/mol
MDL: MFCD00036015
CAS-Nummer: 17722-17-7
EINECS: 241-720-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR