2',4'-Dichlor-5'-hydroxyacetanilid
Lieferant: BIONET RESEARCH
Synonyme:
N-(2,4-Dichloro-5-hydroxyphenyl)acetamide
4L-304S.500
4L-304S.1
4L-304S.5
4L-304S.10
BIOT4L-304S.500EA
0
EUR
BIOT4L-304S.500
BIOT4L-304S.1
BIOT4L-304S.5
BIOT4L-304S.10
2',4'-Dichlor-5'-hydroxyacetanilid
2',4'-Dichlor-5'-hydroxyacetanilid
Formel:
C₈H₇Cl₂NO₂ Molecular Weight: 220.0527 g/mol |
MDL:
MFCD00186236 CAS-Nummer: 67669-19-6 EINECS: 429-110-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...