4'-Cyanacetanilid

Lieferant: Thermo Scientific

Synonyme: N-(4-Cyanophenyl)acetamide

SPB07621.1 SPB07621.10 SPB07621.50
MAYBSPB07621.1EA 0 EUR
MAYBSPB07621.1 MAYBSPB07621.10 MAYBSPB07621.50
4'-Cyanacetanilid
4'-Cyanacetanilid
Formel: C₉H₈N₂O
Molecular Weight: 160,18 g/mol
MDL: MFCD00001814
CAS-Nummer: 35704-19-9
EINECS: 252-685-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR