2-Cyan-3'-methoxyacetanilid

Lieferant: SIA ENAMINE

EN300-01307 EN30001307.1G
ENAMEN300-01307EA 0 EUR
ENAMEN300-01307 ENAMEN30001307.1G
2-Cyan-3'-methoxyacetanilid
2-Cyan-3'-methoxyacetanilid
Formel: C₁₀H₁₀N₂O₂
CAS-Nummer: 91818-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR