2-Cyan-4'-methoxyacetanilid

Lieferant: BIONET RESEARCH

2D-084.500 2D-084.1 2D-084.5 2D-084.10
BIOT2D-084.500EA 0 EUR
BIOT2D-084.500 BIOT2D-084.1 BIOT2D-084.5 BIOT2D-084.10
2-Cyan-4'-methoxyacetanilid
2-Cyan-4'-methoxyacetanilid
Formel: C₁₀H₁₀N₂O₂
MDL: MFCD00214717
CAS-Nummer: 5382-38-7
EINECS: 226-376-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR