4'-Aminoacetanilid Hydrochlorid

Lieferant: Thermo Scientific

SB00801.1 SB00801.10 SB00801EE
MAYBSB00801.1EA 0 EUR
MAYBSB00801.1 MAYBSB00801.10 MAYBSB00801EE
4'-Aminoacetanilid Hydrochlorid
4'-Aminoacetanilid Hydrochlorid
Formel: C₈H₁₁ClN₂O
MDL: MFCD00050695
CAS-Nummer: 43036-07-3
EINECS: 256-052-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR