4'-Methoxy-2'-nitroacetanilid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Acetamido-3-Nitroanisole, N-(4-Methoxy-2-nitro-phenyl)-acetamide, 4-Methoxy-2-nitroacetanilide, N-(4-Methoxy-2-nitrophenyl)acetamide
L04218.06
L04218.14
L04218.06EA
30.4
EUR
L04218.06
L04218.14
4'-Methoxy-2'-nitroacetanilid ≥98%
4'-Methoxy-2'-nitroacetanilid
Purity: 98+%
CAS No.: 119-81-3
Molecular Formula: C9H10N2O4
Formula Weight: 210.19
Melting Point: 117-118°
Beilstein Reference: 2809678
Formel:
C₉H₁₀N₂O₄ Molecular Weight: 210,19 g/mol Schmelzpunkt: 117…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017018 CAS-Nummer: 119-81-3 EINECS: 204-353-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...