4'-Methoxy-2'-nitroacetanilid

Lieferant: BIONET RESEARCH

Synonyme: 4-Acetamido-3-Nitroanisole, N-(4-Methoxy-2-nitro-phenyl)-acetamide, 4-Methoxy-2-nitroacetanilide, N-(4-Methoxy-2-nitrophenyl)acetamide

1N-711.500 1N-711.1 1N-711.5 1N-711.10
BIOT1N-711.500EA 0 EUR
BIOT1N-711.500 BIOT1N-711.1 BIOT1N-711.5 BIOT1N-711.10
4'-Methoxy-2'-nitroacetanilid
4'-Methoxy-2'-nitroacetanilid
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,19 g/mol
MDL: MFCD00017018
CAS-Nummer: 119-81-3
EINECS: 204-353-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR