Trimethylsilylacetat (Acetoxytrimethylsilan)

Lieferant: ABCR

Synonyme: Trimethylsilyl acetate

AB110789.0100 AB110789.0025
ABCRAB110789.0100EA 0 EUR
ABCRAB110789.0100 ABCRAB110789.0025
Trimethylsilylacetat (Acetoxytrimethylsilan)
Trimethylsilylacetat (Acetoxytrimethylsilan)
Formel: C₅H₁₂O₂Si
Molecular Weight: 132,24 g/mol
MDL: MFCD00008695
CAS-Nummer: 2754-27-0
EINECS: 220-404-2
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR